The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001320602.1 | NP_001307531.1 | 2 - 3 | 319 | 107 |
NM_144770.4 | NP_658983.3 | 2 - 3 | 319 | 107 | |
NR_135310.1 | 2 - 3 | 329 | 107 | ||
NR_135311.1 | 2 - 3 | 329 | 107 | ||
XM_005260997.4 | XP_005261054.1 | 2 - 3 | 342 | 107 | |
XM_017028385.1 | XP_016883874.1 | 2 - 3 | 238 | 107 | |
XM_017028386.1 | XP_016883875.1 | 2 - 3 | 474 | 107 | |
XM_017028387.1 | XP_016883876.1 | 2 - 3 | 481 | 107 | |
GenBank mRNA | AK091331.1 | - | 2 - 3 | 301 | 107 |
AY077695.1 | - | 2 - 3 | 250 | 107 | |
BC030196.1 | - | 2 - 3 | 271 | 107 | |
CB153254.1 | - | 2 - 3 | 329 | 107 | |
CN430785.1 | - | 2 - 3 | 315 | 107 |