The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005618.3 | NP_005609.3 | 2 - 3 | 822 | 71 |
XM_005266934.3 | XP_005266991.1 | 2 - 3 | 1288 | 71 | |
GenBank mRNA | AF196571.1 | - | 2 - 3 | 670 | 71 |
AK314234.1 | - | 2 - 3 | 694 | 71 | |
AY358892.1 | - | 2 - 3 | 389 | 71 | |
DR000029.1 | - | 2 - 3 | 441 | 71 | |
EU927387.1 | - | 2 - 3 | 513 | 71 |