The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005618.3 | NP_005609.3 | 3 - 4 | 885 | 61 |
XM_005266934.3 | XP_005266991.1 | 3 - 4 | 1351 | 61 | |
GenBank mRNA | AF196571.1 | - | 3 - 4 | 733 | 61 |
AK314234.1 | - | 3 - 4 | 757 | 61 | |
AY358892.1 | - | 3 - 4 | 452 | 61 | |
DR000029.1 | - | 3 - 4 | 504 | 61 | |
EU927387.1 | - | 3 - 4 | 576 | 61 |