The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018489.2 | NP_060959.2 | 25 - 26 | 8942 | 63 |
XM_006711450.3 | XP_006711513.1 | 25 - 26 | 8633 | 63 | |
XM_006711451.3 | XP_006711514.1 | 26 - 27 | 8720 | 63 | |
XM_006711452.3 | XP_006711515.1 | 24 - 25 | 8762 | 63 | |
XM_011509770.2 | XP_011508072.1 | 24 - 25 | 4313 | 63 | |
XM_017001784.1 | XP_016857273.1 | 25 - 26 | 8719 | 63 | |
XM_017001785.1 | XP_016857274.1 | 25 - 26 | 8649 | 63 | |
XM_017001788.1 | XP_016857277.1 | 22 - 23 | 3437 | 63 | |
GenBank mRNA | AF257305.1 | - | 25 - 26 | 8786 | 63 |
AK304873.1 | - | 6 - 7 | 752 | 63 |