The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001313998.1 | NP_001300927.1 | 4 - 5 | 424 | 67 |
NM_001313999.1 | NP_001300928.1 | 3 - 4 | 262 | 67 | |
NM_003766.4 | NP_003757.1 | 4 - 5 | 446 | 67 | |
XM_005257759.2 | XP_005257816.1 | 4 - 5 | 368 | 67 | |
XM_005257760.3 | XP_005257817.1 | 4 - 5 | 368 | 67 | |
XM_011525421.2 | XP_011523723.1 | 4 - 5 | 368 | 67 | |
XM_017025262.1 | XP_016880751.1 | 4 - 5 | 441 | 67 | |
XM_017025263.1 | XP_016880752.1 | 4 - 5 | 441 | 67 | |
XM_017025264.1 | XP_016880753.1 | 4 - 5 | 441 | 67 | |
GenBank mRNA | AF077301.1 | - | 3 - 4 | 260 | 67 |
AF139131.1 | - | 4 - 5 | 380 | 67 | |
AK223411.1 | - | 4 - 5 | 388 | 67 | |
AK312651.1 | - | 4 - 5 | 371 | 67 | |
BC010276.1 | - | 4 - 5 | 393 | 67 | |
DC296896.1 | - | 4 - 5 | 423 | 67 | |
DC354547.1 | - | 4 - 5 | 446 | 67 | |
KC776730.1 | - | 3 - 4 | 260 | 67 |