Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SRRM1
Entrez Gene ID
10250
Gene Name
serine and arginine repetitive matrix 1
Gene Aliases
160-KD, POP101, SRM160
Chromosome Location
Chr.1: 24643012 - 24673281 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001303448.1 NP_001290377.1 13 - 14 1893 90
NM_001303449.1 NP_001290378.1 13 - 14 1822 90
XM_005245717.2 XP_005245774.1 13 - 14 1809 90
XM_005245718.2 XP_005245775.1 13 - 14 1806 90
XM_005245720.2 XP_005245777.1 13 - 14 1800 90
XM_005245722.2 XP_005245779.1 13 - 14 1719 90
XM_011540478.2 XP_011538780.1 13 - 14 1823 90
XM_011540479.1 XP_011538781.1 13 - 14 3770 90
XM_011540480.1 XP_011538782.1 14 - 15 3078 90
XM_011540481.1 XP_011538783.1 14 - 15 2120 90
XM_011540482.1 XP_011538784.1 13 - 14 3764 90
XM_011540483.1 XP_011538785.1 13 - 14 3680 90
XM_017000011.1 XP_016855500.1 13 - 14 1820 90
XM_017000012.1 XP_016855501.1 13 - 14 1814 90
XM_017000016.1 XP_016855505.1 13 - 14 1755 90
XM_017000017.1 XP_016855506.1 13 - 14 1730 90
XM_017000020.1 XP_016855509.1 13 - 14 1716 90
XM_017000021.1 XP_016855510.1 13 - 14 1713 90
XM_017000023.1 XP_016855512.1 13 - 14 3767 90
XM_017000024.1 XP_016855513.1 13 - 14 3761 90
XM_017000030.1 XP_016855519.1 13 - 14 3677 90
XM_017000031.1 XP_016855520.1 14 - 15 2027 90
XM_017000032.1 XP_016855521.1 13 - 14 3674 90
GenBank mRNA AK293576.1 - 11 - 12 1316 90
AK302498.1 - 13 - 14 1712 90
AK307419.1 - 13 - 14 1822 90
AK307568.1 - 10 - 11 1533 90
KU178513.1 - 13 - 14 1675 90

View Full Product Details