Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DDX51
Entrez Gene ID
317781
Gene Name
DEAD-box helicase 51
Gene Aliases
-
Chromosome Location
Chr.12: 132136594 - 132144335 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_175066.3 NP_778236.2 6 - 7 1038 90
XM_011538256.1 XP_011536558.1 6 - 7 1038 90
GenBank mRNA AB593157.1 - 6 - 7 1058 90
AK097078.1 - 6 - 7 1038 90
BC029279.1 - 6 - 7 1018 90
BC040185.2 - 6 - 7 1011 90

View Full Product Details