The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014553.2 | NP_055368.1 | 14 - 15 | 1485 | 96 |
XM_017003902.1 | XP_016859391.1 | 13 - 14 | 1350 | 96 | |
XM_017003903.1 | XP_016859392.1 | 9 - 10 | 792 | 96 | |
GenBank mRNA | AF198488.1 | - | 14 - 15 | 1482 | 96 |
AK311692.1 | - | 2 - 3 | 115 | 96 | |
BC064698.1 | - | 14 - 15 | 1485 | 96 |