The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000444.5 | NP_000435.3 | 18 - 19 | 2463 | 133 |
NM_001282754.1 | NP_001269683.1 | 18 - 19 | 2463 | 133 | |
XM_011545533.1 | XP_011543835.1 | 14 - 15 | 1272 | 133 | |
XM_011545536.2 | XP_011543838.1 | 10 - 11 | 1305 | 133 | |
XM_017029579.1 | XP_016885068.1 | 14 - 15 | 1273 | 133 | |
GenBank mRNA | AD000712.1 | - | 18 - 19 | 2060 | 133 |
AK298030.1 | - | 18 - 19 | 2009 | 133 | |
AK301528.1 | - | 17 - 18 | 2199 | 133 | |
AK304551.1 | - | 11 - 12 | 1300 | 133 | |
BC105057.1 | - | 18 - 19 | 1985 | 133 | |
BC105059.1 | - | 18 - 19 | 1985 | 133 | |
U75645.1 | - | 18 - 19 | 1977 | 133 | |
U82970.1 | - | 18 - 19 | 2504 | 133 | |
U87284.1 | - | 18 - 19 | 2102 | 133 |