The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000444.5 | NP_000435.3 | 2 - 3 | 748 | 140 |
NM_001282754.1 | NP_001269683.1 | 2 - 3 | 748 | 140 | |
GenBank mRNA | AD000712.1 | - | 2 - 3 | 345 | 140 |
AK298030.1 | - | 2 - 3 | 294 | 140 | |
AK301528.1 | - | 1 - 2 | 484 | 140 | |
BC105057.1 | - | 2 - 3 | 270 | 140 | |
BC105059.1 | - | 2 - 3 | 270 | 140 | |
U60475.1 | - | 2 - 3 | 179 | 140 | |
U75645.1 | - | 2 - 3 | 262 | 140 | |
U82970.1 | - | 2 - 3 | 789 | 140 | |
U87284.1 | - | 2 - 3 | 387 | 140 |