Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SULF1
Entrez Gene ID
23213
Gene Name
sulfatase 1
Gene Aliases
SULF-1
Chromosome Location
Chr.8: 69466624 - 69660912 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001128204.1 NP_001121676.1 3 - 4 497 128
NM_001128205.1 NP_001121677.1 4 - 5 665 128
NM_001128206.1 NP_001121678.1 3 - 4 503 128
NM_015170.2 NP_055985.2 4 - 5 659 128
XM_006716438.2 XP_006716501.1 4 - 5 697 128
XM_006716439.2 XP_006716502.1 3 - 4 503 128
XM_006716440.3 XP_006716503.1 4 - 5 659 128
XM_011517494.1 XP_011515796.1 3 - 4 408 128
XM_011517495.1 XP_011515797.1 4 - 5 697 128
XM_017013250.1 XP_016868739.1 4 - 5 697 128
GenBank mRNA AF545571.1 - 4 - 5 648 128
AK055607.1 - 3 - 4 449 128
AK307951.1 - 3 - 4 276 128
AY101175.1 - 3 - 4 450 128
BX571746.1 - 4 - 5 610 128
DA726387.1 - 3 - 4 503 128

View Full Product Details