Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CAPN11
Entrez Gene ID
11131
Gene Name
calpain 11
Gene Aliases
calpain11
Chromosome Location
Chr.6: 44158811 - 44184402 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_007058.3 NP_008989.2 3 - 4 375 109
XM_006714984.1 XP_006715047.1 3 - 4 378 109
XM_006714985.1 XP_006715048.1 3 - 4 378 109
XM_006714987.1 XP_006715050.1 3 - 4 378 109
XM_011514274.1 XP_011512576.1 3 - 4 459 109
XM_011514275.1 XP_011512577.1 4 - 5 501 109
GenBank mRNA AJ242832.1 - 2 - 3 329 109
AK314054.1 - 3 - 4 355 109
BC033733.1 - 3 - 4 346 109
BG721593.1 - 3 - 4 380 109
DB052011.1 - 4 - 5 465 109

View Full Product Details