Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SLC22A7
Entrez Gene ID
10864
Gene Name
solute carrier family 22 member 7
Gene Aliases
NLT, OAT2
Chromosome Location
Chr.6: 43295694 - 43305538 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_006672.3 NP_006663.2 2 - 3 596 80
NM_153320.2 NP_696961.2 3 - 4 602 80
XM_006714970.3 XP_006715033.1 3 - 4 602 80
XM_006714971.3 XP_006715034.1 2 - 3 596 80
XM_011514256.2 XP_011512558.1 3 - 4 779 80
XM_011514257.2 XP_011512559.1 3 - 4 779 80
XM_011514259.1 XP_011512561.1 3 - 4 779 80
XM_011514261.2 XP_011512563.1 4 - 5 544 80
XM_011514262.2 XP_011512564.1 3 - 4 779 80
XM_011514263.2 XP_011512565.1 3 - 4 355 80
XM_017010198.1 XP_016865687.1 3 - 4 779 80
XM_017010200.1 XP_016865689.1 4 - 5 584 80
XM_017010201.1 XP_016865690.1 4 - 5 554 80
GenBank mRNA AF097518.1 - 3 - 4 598 80
AF210455.1 - 2 - 3 497 80
AK290796.1 - 2 - 3 515 80
AK290806.1 - 2 - 3 574 80
AK309570.1 - 2 - 3 558 80
AY050498.1 - 3 - 4 503 80
BC017963.1 - 2 - 3 809 80
BC033805.1 - 3 - 4 523 80

View Full Product Details