Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SLC22A7
Entrez Gene ID
10864
Gene Name
solute carrier family 22 member 7
Gene Aliases
NLT, OAT2
Chromosome Location
Chr.6: 43295694 - 43305538 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_006672.3 NP_006663.2 5 - 6 1048 93
NM_153320.2 NP_696961.2 6 - 7 1054 93
XM_006714970.3 XP_006715033.1 6 - 7 1063 93
XM_006714971.3 XP_006715034.1 5 - 6 1057 93
XM_011514256.2 XP_011512558.1 6 - 7 1240 93
XM_011514257.2 XP_011512559.1 6 - 7 1231 93
XM_011514259.1 XP_011512561.1 6 - 7 1240 93
XM_011514261.2 XP_011512563.1 7 - 8 1005 93
XM_011514263.2 XP_011512565.1 6 - 7 807 93
XM_017010198.1 XP_016865687.1 6 - 7 1240 93
XM_017010199.1 XP_016865688.1 5 - 6 1081 93
XM_017010200.1 XP_016865689.1 7 - 8 1045 93
XM_017010201.1 XP_016865690.1 7 - 8 1015 93
GenBank mRNA AF097518.1 - 6 - 7 1050 93
AF210455.1 - 5 - 6 949 93
AK290796.1 - 5 - 6 967 93
AK290806.1 - 5 - 6 1026 93
AK309570.1 - 5 - 6 1019 93
AY050498.1 - 6 - 7 955 93
BC033805.1 - 6 - 7 975 93

View Full Product Details