The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003047.4 | NP_003038.2 | 1 - 2 | 1148 | 64 |
GenBank mRNA | AF141350.1 | - | 1 - 2 | 395 | 64 |
AF141351.1 | - | 1 - 2 | 395 | 64 | |
AF141352.1 | - | 1 - 2 | 395 | 64 | |
AF141353.1 | - | 1 - 2 | 395 | 64 | |
AF141354.1 | - | 1 - 2 | 395 | 64 | |
AF141355.1 | - | 1 - 2 | 395 | 64 | |
AF141356.1 | - | 1 - 2 | 395 | 64 | |
AF141357.1 | - | 1 - 2 | 395 | 64 | |
AF141358.1 | - | 1 - 2 | 395 | 64 | |
AF141359.1 | - | 1 - 2 | 395 | 64 | |
AF146430.1 | - | 1 - 2 | 395 | 64 | |
AF146431.1 | - | 1 - 2 | 395 | 64 | |
AF146432.1 | - | 1 - 2 | 395 | 64 | |
AF146433.1 | - | 1 - 2 | 395 | 64 | |
AF146434.1 | - | 1 - 2 | 395 | 64 | |
AF146435.1 | - | 1 - 2 | 395 | 64 | |
AF146436.1 | - | 1 - 2 | 395 | 64 | |
AF146437.1 | - | 1 - 2 | 395 | 64 | |
AF146438.1 | - | 1 - 2 | 395 | 64 | |
AF146439.1 | - | 1 - 2 | 395 | 64 | |
AK314190.1 | - | 1 - 2 | 543 | 64 | |
BC012121.1 | - | 1 - 2 | 616 | 64 | |
M81768.1 | - | 1 - 2 | 405 | 64 | |
M96066.1 | - | 1 - 2 | 929 | 64 | |
S68616.1 | - | 1 - 2 | 929 | 64 |