The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003047.4 | NP_003038.2 | 7 - 8 | 2444 | 72 |
NR_046474.1 | 6 - 7 | 1983 | 72 | ||
XM_011542021.2 | XP_011540323.1 | 8 - 9 | 1522 | 72 | |
GenBank mRNA | AF141350.1 | - | 7 - 8 | 1691 | 72 |
AF141351.1 | - | 7 - 8 | 1691 | 72 | |
AF141352.1 | - | 7 - 8 | 1691 | 72 | |
AF141353.1 | - | 7 - 8 | 1691 | 72 | |
AF141354.1 | - | 7 - 8 | 1691 | 72 | |
AF141355.1 | - | 7 - 8 | 1691 | 72 | |
AF141356.1 | - | 7 - 8 | 1691 | 72 | |
AF141357.1 | - | 7 - 8 | 1691 | 72 | |
AF141358.1 | - | 7 - 8 | 1691 | 72 | |
AF141359.1 | - | 7 - 8 | 1691 | 72 | |
AF146430.1 | - | 7 - 8 | 1691 | 72 | |
AF146431.1 | - | 7 - 8 | 1691 | 72 | |
AF146432.1 | - | 7 - 8 | 1691 | 72 | |
AF146433.1 | - | 7 - 8 | 1691 | 72 | |
AF146434.1 | - | 7 - 8 | 1691 | 72 | |
AF146435.1 | - | 7 - 8 | 1691 | 72 | |
AF146436.1 | - | 7 - 8 | 1691 | 72 | |
AF146437.1 | - | 7 - 8 | 1691 | 72 | |
AF146438.1 | - | 7 - 8 | 1691 | 72 | |
AF146439.1 | - | 7 - 8 | 1691 | 72 | |
AK124218.1 | - | 2 - 3 | 873 | 72 | |
AK300432.1 | - | 6 - 7 | 1335 | 72 | |
AK314190.1 | - | 7 - 8 | 1839 | 72 | |
M81768.1 | - | 7 - 8 | 1701 | 72 | |
M96066.1 | - | 7 - 8 | 2225 | 72 | |
M96067.1 | - | 3 - 4 | 181 | 72 | |
S68616.1 | - | 7 - 8 | 2225 | 72 |