The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_153252.4 | NP_694984.4 | 36 - 37 | 4337 | 97 |
XM_005262113.3 | XP_005262170.1 | 35 - 36 | 4189 | 97 | |
XM_017029384.1 | XP_016884873.1 | 32 - 33 | 3867 | 97 | |
GenBank mRNA | AK095887.1 | - | 19 - 20 | 2210 | 97 |
AY497046.1 | - | 36 - 37 | 4337 | 97 | |
AY497047.1 | - | 33 - 34 | 4269 | 97 | |
AY497048.1 | - | 36 - 37 | 4360 | 97 | |
AY497049.1 | - | 35 - 36 | 4330 | 97 | |
AY497050.1 | - | 36 - 37 | 4310 | 97 | |
AY497051.1 | - | 33 - 34 | 3897 | 97 | |
AY497052.1 | - | 38 - 39 | 4496 | 97 | |
AY497053.1 | - | 34 - 35 | 4085 | 97 | |
AY497054.1 | - | 33 - 34 | 3986 | 97 | |
AY497055.1 | - | 36 - 37 | 4271 | 97 | |
AY497056.1 | - | 33 - 34 | 4242 | 97 | |
AY497057.1 | - | 32 - 33 | 4141 | 97 | |
AY497058.1 | - | 32 - 33 | 3980 | 97 | |
AY497059.1 | - | 34 - 35 | 4351 | 97 | |
AY497060.1 | - | 30 - 31 | 3918 | 97 | |
BC039508.2 | - | 8 - 9 | 810 | 97 |