The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_153252.4 | NP_694984.4 | 20 - 21 | 2586 | 104 |
XM_017029384.1 | XP_016884873.1 | 16 - 17 | 2116 | 104 | |
XM_017029385.1 | XP_016884874.1 | 20 - 21 | 2588 | 104 | |
GenBank mRNA | AK095887.1 | - | 3 - 4 | 459 | 104 |
AY497046.1 | - | 20 - 21 | 2586 | 104 | |
AY497047.1 | - | 17 - 18 | 2518 | 104 | |
AY497048.1 | - | 20 - 21 | 2609 | 104 | |
AY497049.1 | - | 19 - 20 | 2579 | 104 | |
AY497050.1 | - | 20 - 21 | 2559 | 104 | |
AY497051.1 | - | 17 - 18 | 2146 | 104 | |
AY497052.1 | - | 22 - 23 | 2745 | 104 | |
AY497053.1 | - | 18 - 19 | 2334 | 104 | |
AY497054.1 | - | 17 - 18 | 2235 | 104 | |
AY497055.1 | - | 20 - 21 | 2520 | 104 | |
AY497056.1 | - | 17 - 18 | 2491 | 104 | |
AY497057.1 | - | 16 - 17 | 2390 | 104 | |
AY497058.1 | - | 16 - 17 | 2229 | 104 | |
AY497059.1 | - | 18 - 19 | 2600 | 104 | |
AY497060.1 | - | 14 - 15 | 2167 | 104 |