The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_153252.4 | NP_694984.4 | 21 - 22 | 2740 | 69 |
XM_017029384.1 | XP_016884873.1 | 17 - 18 | 2270 | 69 | |
GenBank mRNA | AK095887.1 | - | 4 - 5 | 613 | 69 |
AY497046.1 | - | 21 - 22 | 2740 | 69 | |
AY497047.1 | - | 18 - 19 | 2672 | 69 | |
AY497048.1 | - | 21 - 22 | 2763 | 69 | |
AY497049.1 | - | 20 - 21 | 2733 | 69 | |
AY497050.1 | - | 21 - 22 | 2713 | 69 | |
AY497051.1 | - | 18 - 19 | 2300 | 69 | |
AY497052.1 | - | 23 - 24 | 2899 | 69 | |
AY497053.1 | - | 19 - 20 | 2488 | 69 | |
AY497054.1 | - | 18 - 19 | 2389 | 69 | |
AY497055.1 | - | 21 - 22 | 2674 | 69 | |
AY497056.1 | - | 18 - 19 | 2645 | 69 | |
AY497057.1 | - | 17 - 18 | 2544 | 69 | |
AY497058.1 | - | 17 - 18 | 2383 | 69 | |
AY497059.1 | - | 19 - 20 | 2754 | 69 | |
AY497060.1 | - | 15 - 16 | 2321 | 69 |