The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001278579.1 | NP_001265508.1 | 5 - 6 | 804 | 110 |
NM_001278580.1 | NP_001265509.1 | 4 - 5 | 607 | 110 | |
NM_001616.4 | NP_001607.1 | 4 - 5 | 693 | 110 | |
GenBank mRNA | AK301629.1 | - | 4 - 5 | 607 | 110 |
AK314124.1 | - | 4 - 5 | 883 | 110 | |
BC067417.1 | - | 4 - 5 | 621 | 110 | |
BC067418.1 | - | 4 - 5 | 621 | 110 | |
BC069707.1 | - | 4 - 5 | 688 | 110 | |
D31770.1 | - | 4 - 5 | 698 | 110 | |
M93415.1 | - | 5 - 6 | 708 | 110 | |
X62381.1 | - | 4 - 5 | 680 | 110 | |
X63128.1 | - | 4 - 5 | 550 | 110 |