The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012259.2 | NP_036391.1 | 3 - 4 | 439 | 90 |
XM_017010627.1 | XP_016866116.1 | 3 - 4 | 1823 | 90 | |
XM_017010628.1 | XP_016866117.1 | 4 - 5 | 1521 | 90 | |
XM_017010629.1 | XP_016866118.1 | 3 - 4 | 300 | 90 | |
GenBank mRNA | AB044755.1 | - | 3 - 4 | 344 | 90 |
AF173901.1 | - | 3 - 4 | 407 | 90 | |
AF232238.1 | - | 3 - 4 | 358 | 90 | |
AF237949.1 | - | 3 - 4 | 315 | 90 | |
AF311884.1 | - | 3 - 4 | 248 | 90 | |
AJ249545.1 | - | 3 - 4 | 298 | 90 | |
AK289943.1 | - | 3 - 4 | 357 | 90 | |
AK303890.1 | - | 3 - 4 | 261 | 90 | |
BC007707.2 | - | 3 - 4 | 337 | 90 | |
BT020067.1 | - | 3 - 4 | 248 | 90 | |
BT020068.1 | - | 3 - 4 | 248 | 90 |