Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HEY2
Entrez Gene ID
23493
Gene Name
hes related family bHLH transcription factor with YRPW motif 2
Gene Aliases
CHF1, GRIDLOCK, GRL, HERP1, HESR2, HRT2, bHLHb32
Chromosome Location
Chr.6: 125747592 - 125762243 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_012259.2 NP_036391.1 4 - 5 509 96
XM_017010627.1 XP_016866116.1 4 - 5 1893 96
XM_017010628.1 XP_016866117.1 5 - 6 1591 96
XM_017010629.1 XP_016866118.1 4 - 5 370 96
GenBank mRNA AB044755.1 - 4 - 5 414 96
AF173901.1 - 4 - 5 477 96
AF232238.1 - 4 - 5 428 96
AF237949.1 - 4 - 5 385 96
AF311884.1 - 4 - 5 318 96
AJ249545.1 - 4 - 5 368 96
AK289943.1 - 4 - 5 427 96
AK303890.1 - 4 - 5 331 96
BC007707.2 - 4 - 5 407 96
BT020067.1 - 4 - 5 318 96
BT020068.1 - 4 - 5 318 96

View Full Product Details