The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001314030.1 | NP_001300959.1 | 11 - 12 | 2396 | 120 |
NM_012284.2 | NP_036416.1 | 11 - 12 | 2400 | 120 | |
XM_011538085.2 | XP_011536387.1 | 11 - 12 | 3016 | 120 | |
XM_011538086.2 | XP_011536388.1 | 11 - 12 | 3012 | 120 | |
XM_017019096.1 | XP_016874585.1 | 10 - 11 | 2782 | 120 | |
XM_017019097.1 | XP_016874586.1 | 11 - 12 | 2020 | 120 | |
GenBank mRNA | AB022696.1 | - | 11 - 12 | 2175 | 120 |
AB033108.1 | - | 11 - 12 | 2400 | 120 | |
BC150602.1 | - | 11 - 12 | 2281 | 120 |