The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014400.2 | NP_055215.2 | 2 - 3 | 319 | 102 |
GenBank mRNA | AF082889.1 | - | 2 - 3 | 296 | 102 |
AJ223603.2 | - | 2 - 3 | 247 | 102 | |
AK075464.1 | - | 2 - 3 | 248 | 102 | |
AK290905.1 | - | 2 - 3 | 248 | 102 | |
AK314773.1 | - | 2 - 3 | 248 | 102 | |
AL832153.1 | - | 2 - 3 | 249 | 102 | |
AY359006.1 | - | 2 - 3 | 226 | 102 | |
BC039167.1 | - | 2 - 3 | 233 | 102 | |
BC041061.1 | - | 2 - 3 | 228 | 102 | |
BP367221.1 | - | 2 - 3 | 293 | 102 | |
DA837818.1 | - | 2 - 3 | 319 | 102 |