The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001005367.2 | NP_001005367.1 | 10 - 11 | 1242 | 84 |
NM_001201461.1 | NP_001188390.1 | 10 - 11 | 1242 | 84 | |
NM_020659.3 | NP_065710.1 | 10 - 11 | 1242 | 84 | |
XM_017027025.1 | XP_016882514.1 | 10 - 11 | 1086 | 84 | |
XM_017027026.1 | XP_016882515.1 | 10 - 11 | 1086 | 84 | |
GenBank mRNA | AB162930.1 | - | 10 - 11 | 1125 | 84 |
AB188496.1 | - | 10 - 11 | 1242 | 84 | |
AF177909.1 | - | 10 - 11 | 1156 | 84 | |
AK289468.1 | - | 10 - 11 | 1205 | 84 | |
AK293468.1 | - | 11 - 12 | 1273 | 84 | |
BC011347.1 | - | 10 - 11 | 1147 | 84 | |
BC019358.1 | - | 7 - 8 | 575 | 84 |