Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SYMPK
Entrez Gene ID
8189
Gene Name
symplekin
Gene Aliases
SPK, SYM
Chromosome Location
Chr.19: 45815442 - 45863290 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_004819.2 NP_004810.2 9 - 10 1333 72
XM_005259286.1 XP_005259343.1 9 - 10 1286 72
XM_011527354.1 XP_011525656.1 10 - 11 1336 72
GenBank mRNA AB209024.1 - 9 - 10 3581 72
AB210010.1 - 9 - 10 1178 72
AK300252.1 - 9 - 10 1187 72
BC006536.2 - 8 - 9 1040 72
BC006567.2 - 8 - 9 1040 72
BC030214.1 - 9 - 10 1175 72
BX649165.1 - 9 - 10 1188 72
U49240.1 - 4 - 5 802 72
Y10931.1 - 9 - 10 1151 72

View Full Product Details