The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_021255.2 | NP_067078.1 | 2 - 3 | 492 | 99 |
XM_005267890.4 | XP_005267947.1 | 2 - 3 | 537 | 99 | |
XM_011536990.2 | XP_011535292.1 | 3 - 4 | 202 | 99 | |
XM_011536992.2 | XP_011535294.1 | 2 - 3 | 255 | 99 | |
XM_017021478.1 | XP_016876967.1 | 2 - 3 | 884 | 99 | |
XM_017021479.1 | XP_016876968.1 | 3 - 4 | 9434 | 99 | |
GenBank mRNA | AF302502.1 | - | 2 - 3 | 382 | 99 |
AK315727.1 | - | 2 - 3 | 454 | 99 | |
BC009476.2 | - | 2 - 3 | 492 | 99 |