Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NUP43
Entrez Gene ID
348995
Gene Name
nucleoporin 43
Gene Aliases
bA350J20.1, p42
Chromosome Location
Chr.6: 149724315 - 149746572 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_198887.2 NP_942590.1 4 - 5 579 81
NR_104456.1 4 - 5 579 81
XM_005266960.4 XP_005267017.1 4 - 5 529 81
XM_005266961.3 XP_005267018.1 4 - 5 529 81
XM_005266962.3 XP_005267019.1 4 - 5 529 81
XM_011535798.2 XP_011534100.1 4 - 5 529 81
XM_011535799.2 XP_011534101.1 4 - 5 529 81
GenBank mRNA AF514997.1 - 4 - 5 512 81
AK074311.1 - - 534 81
BC040121.1 - 2 - 3 224 81
BC065028.1 - 4 - 5 505 81
BX647097.1 - 4 - 5 529 81
CB989591.1 - 4 - 5 576 81

View Full Product Details