The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000495.4 | NP_000486.1 | 27 - 28 | 2428 | 126 |
NM_033380.2 | NP_203699.1 | 27 - 28 | 2428 | 126 | |
XM_011530849.2 | XP_011529151.2 | 28 - 29 | 2176 | 126 | |
XM_017029259.1 | XP_016884748.1 | 28 - 29 | 2176 | 126 | |
XM_017029260.1 | XP_016884749.1 | 28 - 29 | 2176 | 126 | |
XM_017029261.1 | XP_016884750.1 | 28 - 29 | 2176 | 126 | |
XM_017029262.1 | XP_016884751.1 | 28 - 29 | 2176 | 126 | |
XM_017029263.1 | XP_016884752.1 | 12 - 13 | 1254 | 126 | |
GenBank mRNA | BC035387.1 | - | 11 - 12 | 1603 | 126 |
BC151846.1 | - | 27 - 28 | 2390 | 126 | |
KM282618.1 | - | 27 - 28 | 2428 | 126 | |
M58526.1 | - | 24 - 25 | 1904 | 126 | |
M90464.1 | - | 27 - 28 | 2348 | 126 |