The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000495.4 | NP_000486.1 | 28 - 29 | 2523 | 92 |
NM_033380.2 | NP_203699.1 | 28 - 29 | 2523 | 92 | |
XM_011530849.2 | XP_011529151.2 | 29 - 30 | 2271 | 92 | |
XM_017029259.1 | XP_016884748.1 | 29 - 30 | 2271 | 92 | |
XM_017029260.1 | XP_016884749.1 | 29 - 30 | 2271 | 92 | |
XM_017029261.1 | XP_016884750.1 | 29 - 30 | 2271 | 92 | |
XM_017029262.1 | XP_016884751.1 | 29 - 30 | 2271 | 92 | |
XM_017029263.1 | XP_016884752.1 | 13 - 14 | 1349 | 92 | |
GenBank mRNA | BC035387.1 | - | 12 - 13 | 1698 | 92 |
BC151846.1 | - | 28 - 29 | 2485 | 92 | |
KM282618.1 | - | 28 - 29 | 2523 | 92 | |
M58526.1 | - | 25 - 26 | 1999 | 92 | |
M90464.1 | - | 28 - 29 | 2443 | 92 |