Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
COL4A5
Entrez Gene ID
1287
Gene Name
collagen type IV alpha 5 chain
Gene Aliases
ASLN, ATS, CA54
Chromosome Location
Chr.X: 108439844 - 108697545 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000495.4 NP_000486.1 46 - 47 4575 123
NM_033380.2 NP_203699.1 48 - 49 4593 123
XM_011530849.2 XP_011529151.2 49 - 50 4341 123
XM_017029259.1 XP_016884748.1 48 - 49 4332 123
XM_017029260.1 XP_016884749.1 47 - 48 4323 123
XM_017029263.1 XP_016884752.1 33 - 34 3419 123
GenBank mRNA AK307900.1 - 6 - 7 805 123
BC151846.1 - 48 - 49 4555 123
KM282618.1 - 49 - 50 4566 123
M31115.1 - 15 - 16 1554 123
M58526.1 - 43 - 44 4051 123

View Full Product Details