Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
COL4A5
Entrez Gene ID
1287
Gene Name
collagen type IV alpha 5 chain
Gene Aliases
ASLN, ATS, CA54
Chromosome Location
Chr.X: 108439844 - 108697545 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000495.4 NP_000486.1 49 - 50 5084 86
NM_033380.2 NP_203699.1 51 - 52 5102 86
XM_011530849.2 XP_011529151.2 52 - 53 4850 86
XM_017029259.1 XP_016884748.1 51 - 52 4841 86
XM_017029260.1 XP_016884749.1 50 - 51 4832 86
XM_017029263.1 XP_016884752.1 36 - 37 3928 86
GenBank mRNA AK294066.1 - 2 - 3 196 86
AK307900.1 - 9 - 10 1314 86
BC151846.1 - 51 - 52 5064 86
KM282618.1 - 52 - 53 5075 86
M31115.1 - 18 - 19 2063 86
M58526.1 - 46 - 47 4560 86

View Full Product Details