The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020244.2 | NP_064629.2 | 2 - 3 | 638 | 74 |
XM_011538574.1 | XP_011536876.1 | 2 - 3 | 605 | 74 | |
XM_011538575.1 | XP_011536877.1 | 1 - 2 | 142 | 74 | |
GenBank mRNA | AF047431.1 | - | 2 - 3 | 209 | 74 |
AF111803.1 | - | 1 - 2 | 73 | 74 | |
AF195623.1 | - | 2 - 3 | 585 | 74 | |
AF195624.1 | - | 2 - 3 | 585 | 74 | |
AK075211.1 | - | 2 - 3 | 605 | 74 | |
AK093067.1 | - | 3 - 4 | 497 | 74 | |
AK226127.1 | - | 2 - 3 | 312 | 74 | |
BC020819.1 | - | 2 - 3 | 592 | 74 | |
BC050429.1 | - | 2 - 3 | 631 | 74 |