Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CDKL1
Entrez Gene ID
8814
Gene Name
cyclin dependent kinase like 1
Gene Aliases
KKIALRE, P42
Chromosome Location
Chr.14: 50329404 - 50397404 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001282236.1 NP_001269165.1 3 - 4 942 80
NM_004196.4 NP_004187.2 2 - 3 350 80
XM_005268157.2 XP_005268214.1 3 - 4 942 80
XM_005268159.2 XP_005268216.1 3 - 4 942 80
XM_011537276.1 XP_011535578.1 3 - 4 352 80
XM_017021729.1 XP_016877218.1 3 - 4 834 80
XM_017021730.1 XP_016877219.1 3 - 4 834 80
XM_017021731.1 XP_016877220.1 3 - 4 843 80
XM_017021732.1 XP_016877221.1 3 - 4 834 80
GenBank mRNA AK308732.1 - 3 - 4 937 80
BC104977.1 - 2 - 3 325 80
X66358.1 - 2 - 3 510 80

View Full Product Details