The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001127713.1 | NP_001121185.1 | 5 - 6 | 940 | 78 |
NM_015915.4 | NP_056999.2 | 4 - 5 | 756 | 78 | |
NM_181598.3 | NP_853629.2 | 4 - 5 | 756 | 78 | |
GenBank mRNA | AF131801.1 | - | 3 - 4 | 280 | 78 |
AF444143.1 | - | 4 - 5 | 687 | 78 | |
AK223436.1 | - | 4 - 5 | 669 | 78 | |
AK290185.1 | - | 4 - 5 | 669 | 78 | |
AL833591.1 | - | 5 - 6 | 724 | 78 | |
AY032844.1 | - | 4 - 5 | 684 | 78 | |
BC010708.2 | - | 4 - 5 | 659 | 78 | |
CR457153.1 | - | 3 - 4 | 266 | 78 | |
KU178621.1 | - | 4 - 5 | 515 | 78 | |
KU178622.1 | - | 4 - 5 | 515 | 78 | |
KU178623.1 | - | 4 - 5 | 515 | 78 |