Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ATL1
Entrez Gene ID
51062
Gene Name
atlastin GTPase 1
Gene Aliases
AD-FSP, FSP1, GBP3, HSN1D, SPG3, SPG3A, atlastin1
Chromosome Location
Chr.14: 50533082 - 50633068 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001127713.1 NP_001121185.1 8 - 9 1149 89
NM_015915.4 NP_056999.2 7 - 8 965 89
NM_181598.3 NP_853629.2 7 - 8 965 89
GenBank mRNA AF131801.1 - 6 - 7 489 89
AF444143.1 - 7 - 8 896 89
AK223436.1 - 7 - 8 878 89
AK290185.1 - 7 - 8 878 89
AK312518.1 - 7 - 8 851 89
AL833591.1 - 8 - 9 933 89
AY032844.1 - 7 - 8 893 89
BC010708.2 - 7 - 8 868 89
CR457153.1 - 6 - 7 475 89
KU178621.1 - 7 - 8 724 89
KU178622.1 - 7 - 8 724 89
KU178623.1 - 7 - 8 724 89

View Full Product Details