Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PLEKHJ1
Entrez Gene ID
55111
Gene Name
pleckstrin homology domain containing J1
Gene Aliases
GNRPX
Chromosome Location
Chr.19: 2229952 - 2236724 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018049.2 NP_060519.1 4 - 5 428 108
XM_017026928.1 XP_016882417.1 4 - 5 742 108
GenBank mRNA AK001159.1 - 4 - 5 404 108
AK097723.1 - 5 - 6 1435 108
AK098725.1 - 4 - 5 518 108
BC003084.2 - 4 - 5 349 108
BC008966.2 - 4 - 5 375 108
CR457253.1 - 4 - 5 323 108

View Full Product Details