The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001136139.2 | NP_001129611.1 | 18 - 19 | 2000 | 77 |
XM_006722855.3 | XP_006722918.1 | 19 - 20 | 2203 | 77 | |
XM_006722857.3 | XP_006722920.1 | 19 - 20 | 2201 | 77 | |
XM_017027177.1 | XP_016882666.1 | 19 - 20 | 2213 | 77 | |
XM_017027179.1 | XP_016882668.1 | 19 - 20 | 2201 | 77 | |
XM_017027181.1 | XP_016882670.1 | 19 - 20 | 2125 | 77 | |
XM_017027182.1 | XP_016882671.1 | 19 - 20 | 2115 | 77 | |
GenBank mRNA | AK310998.1 | - | 1 - 2 | 81 | 77 |