Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
KLK7
Entrez Gene ID
5650
Gene Name
kallikrein related peptidase 7
Gene Aliases
PRSS6, SCCE, hK7
Chromosome Location
Chr.19: 50976479 - 50984064 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001207053.1 NP_001193982.1 3 - 4 610 82
NM_001243126.1 NP_001230055.1 3 - 4 593 82
NM_005046.3 NP_005037.1 4 - 5 741 82
NM_139277.2 NP_644806.1 4 - 5 571 82
GenBank mRNA AF411214.1 - 4 - 5 715 82
AF411215.1 - 3 - 4 584 82
AK289660.1 - 4 - 5 594 82
AK295313.1 - 3 - 4 593 82
AY601109.1 - 3 - 4 469 82
AY646152.1 - 2 - 3 325 82
BC032005.1 - 4 - 5 580 82
KF963189.1 - 3 - 4 433 82
KF963190.1 - 3 - 4 434 82
KF963191.1 - 3 - 4 434 82
KF963193.1 - 3 - 4 434 82
L33404.1 - 3 - 4 484 82

View Full Product Details