Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
THOC7
Entrez Gene ID
80145
Gene Name
THO complex 7
Gene Aliases
NIF3L1BP1, fSAP24, hTREX30
Chromosome Location
Chr.3: 63833870 - 63864478 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001285404.1 NP_001272333.1 2 - 3 397 108
NM_025075.3 NP_079351.2 2 - 3 266 108
XM_006713339.3 XP_006713402.1 2 - 3 215 108
GenBank mRNA AI391685.1 - 2 - 3 246 108
AK225110.1 - 2 - 3 154 108
BC020599.2 - 1 - 2 114 108
BC065012.1 - 2 - 3 136 108
BI914906.1 - 2 - 3 397 108
CR457373.1 - 2 - 3 133 108
HY023143.1 - 2 - 3 266 108

View Full Product Details