The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001304549.1 | NP_001291478.1 | 2 - 3 | 515 | 72 |
NM_014209.3 | NP_055024.2 | 2 - 3 | 515 | 72 | |
GenBank mRNA | BC107123.1 | - | 2 - 3 | 113 | 72 |
BC107124.1 | - | 2 - 3 | 113 | 72 | |
BC140746.1 | - | 2 - 3 | 205 | 72 | |
BC144448.1 | - | 2 - 3 | 206 | 72 | |
DB046527.1 | - | 2 - 3 | 507 | 72 |