The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014431.2 | NP_055246.2 | 11 - 12 | 1610 | 76 |
XM_011539637.2 | XP_011537939.1 | 11 - 12 | 2796 | 76 | |
XM_011539638.2 | XP_011537940.1 | 11 - 12 | 1383 | 76 | |
XM_017016072.1 | XP_016871561.1 | 11 - 12 | 1381 | 76 | |
GenBank mRNA | AB033100.2 | - | 11 - 12 | 1608 | 76 |
BC040163.1 | - | 9 - 10 | 1113 | 76 | |
BC136375.1 | - | 11 - 12 | 1492 | 76 | |
BC136376.1 | - | 11 - 12 | 1492 | 76 |