The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014431.2 | NP_055246.2 | 14 - 15 | 2009 | 97 |
XM_011539637.2 | XP_011537939.1 | 14 - 15 | 3195 | 97 | |
XM_011539638.2 | XP_011537940.1 | 14 - 15 | 1782 | 97 | |
XM_017016072.1 | XP_016871561.1 | 14 - 15 | 1780 | 97 | |
GenBank mRNA | AB033100.2 | - | 14 - 15 | 2007 | 97 |
BC040163.1 | - | 12 - 13 | 1512 | 97 | |
BC136375.1 | - | 14 - 15 | 1891 | 97 | |
BC136376.1 | - | 14 - 15 | 1891 | 97 |