The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014431.2 | NP_055246.2 | 4 - 5 | 750 | 66 |
XM_011539637.2 | XP_011537939.1 | 4 - 5 | 1936 | 66 | |
XM_011539638.2 | XP_011537940.1 | 4 - 5 | 523 | 66 | |
XM_017016072.1 | XP_016871561.1 | 4 - 5 | 521 | 66 | |
GenBank mRNA | AB033100.2 | - | 4 - 5 | 748 | 66 |
BC040163.1 | - | 2 - 3 | 253 | 66 | |
BC136375.1 | - | 4 - 5 | 632 | 66 | |
BC136376.1 | - | 4 - 5 | 632 | 66 |