The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020962.2 | NP_066013.1 | 15 - 16 | 2921 | 57 |
XM_011521845.2 | XP_011520147.1 | 15 - 16 | 3101 | 57 | |
XM_011521846.2 | XP_011520148.1 | 15 - 16 | 2728 | 57 | |
XM_011521847.1 | XP_011520149.1 | 15 - 16 | 2788 | 57 | |
XM_017022448.1 | XP_016877937.1 | 15 - 16 | 3106 | 57 | |
GenBank mRNA | AB046848.1 | - | 9 - 10 | 1981 | 57 |
AB052622.1 | - | 15 - 16 | 2921 | 57 |