Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
VWA9
Entrez Gene ID
81556
Gene Name
von Willebrand factor A domain containing 9
Gene Aliases
C15orf44
Chromosome Location
Chr.15: 65578757 - 65611289 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001136043.2 NP_001129515.1 4 - 5 592 96
NM_001207058.1 NP_001193987.1 4 - 5 571 96
NM_001207059.1 NP_001193988.1 3 - 4 451 96
NR_045104.1 4 - 5 735 96
NR_045105.2 4 - 5 989 96
NR_045106.2 4 - 5 896 96
NR_045901.1 3 - 4 627 96
XM_006720708.3 XP_006720771.1 4 - 5 671 96
XM_006720709.3 XP_006720772.1 4 - 5 736 96
XM_006720711.2 XP_006720774.1 3 - 4 595 96
XM_011522101.2 XP_011520403.1 4 - 5 417 96
XM_017022644.1 XP_016878133.1 4 - 5 816 96
GenBank mRNA AK027475.1 - 4 - 5 577 96
AK293207.1 - 3 - 4 404 96
AK296134.1 - 4 - 5 962 96
AK301033.1 - 4 - 5 434 96
AK301222.1 - 3 - 4 580 96
AK301742.1 - 4 - 5 413 96
AL136662.1 - 4 - 5 817 96
BC007991.2 - 4 - 5 562 96
DC298728.1 - 3 - 4 404 96
DC384988.1 - 4 - 5 488 96

View Full Product Details