Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ITGA11
Entrez Gene ID
22801
Gene Name
integrin subunit alpha 11
Gene Aliases
HsT18964
Chromosome Location
Chr.15: 68297433 - 68432312 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001004439.1 NP_001004439.1 9 - 10 1149 82
XM_005254228.3 XP_005254285.1 7 - 8 1371 82
XM_011521363.2 XP_011519665.1 7 - 8 1101 82
GenBank mRNA AF109681.1 - 9 - 10 1134 82
AF111799.1 - 1 - 2 114 82
AF137378.2 - 9 - 10 1152 82
AK095863.1 - 10 - 11 1161 82
AK292445.1 - 9 - 10 1158 82
AK304778.1 - 9 - 10 1159 82
BX496541.1 - 1 - 2 138 82

View Full Product Details