The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001170690.1 | NP_001164161.1 | 10 - 11 | 1281 | 73 |
NM_020974.2 | NP_066025.2 | 10 - 11 | 1281 | 73 | |
XM_005253032.4 | XP_005253089.2 | 10 - 11 | 1277 | 73 | |
XM_005253033.4 | XP_005253090.2 | 10 - 11 | 1277 | 73 | |
XM_005253035.4 | XP_005253092.2 | 10 - 11 | 1277 | 73 | |
XM_005253037.4 | XP_005253094.2 | 10 - 11 | 1277 | 73 | |
XM_011520246.2 | XP_011518548.1 | 10 - 11 | 1277 | 73 | |
XM_011520247.2 | XP_011518549.1 | 10 - 11 | 1277 | 73 | |
XM_011520248.2 | XP_011518550.1 | 10 - 11 | 1277 | 73 | |
GenBank mRNA | AK123039.1 | - | 10 - 11 | 1277 | 73 |
AK131552.1 | - | 10 - 11 | 1275 | 73 | |
BC111690.1 | - | 10 - 11 | 1224 | 73 |