The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001170690.1 | NP_001164161.1 | 13 - 14 | 1780 | 73 |
XM_005253032.4 | XP_005253089.2 | 17 - 18 | 2241 | 73 | |
XM_005253033.4 | XP_005253090.2 | 16 - 17 | 2154 | 73 | |
XM_005253034.4 | XP_005253091.2 | 16 - 17 | 2118 | 73 | |
XM_005253035.4 | XP_005253092.2 | 17 - 18 | 2241 | 73 | |
XM_005253036.4 | XP_005253093.2 | 14 - 15 | 1914 | 73 | |
XM_005253037.4 | XP_005253094.2 | 13 - 14 | 1776 | 73 | |
XM_011520246.2 | XP_011518548.1 | 17 - 18 | 2241 | 73 | |
XM_011520247.2 | XP_011518549.1 | 15 - 16 | 2037 | 73 | |
XM_017018080.1 | XP_016873569.1 | 15 - 16 | 2031 | 73 | |
XM_017018081.1 | XP_016873570.1 | 15 - 16 | 2031 | 73 | |
XM_017018082.1 | XP_016873571.1 | 14 - 15 | 1914 | 73 | |
GenBank mRNA | AK092553.1 | - | 4 - 5 | 605 | 73 |
AK131552.1 | - | 13 - 14 | 1774 | 73 | |
BC111690.1 | - | 13 - 14 | 1723 | 73 |