The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020645.2 | NP_065696.1 | 5 - 6 | 723 | 64 |
GenBank mRNA | AK024750.1 | - | 5 - 6 | 676 | 64 |
AK098694.1 | - | 5 - 6 | 676 | 64 | |
AK296484.1 | - | 4 - 5 | 535 | 64 | |
AY211930.1 | - | 4 - 5 | 420 | 64 | |
BC008835.2 | - | 5 - 6 | 659 | 64 |